Learn More
Chemglass Life Sciences Condenser, "B" Assembly, For Rotary Evaporator, Plastic Coated
SDP
Catalog No. CG132910
Encompass_Preferred
Condenser, "B" Assembly, For Rotary Evaporator, Plastic Coated. Double coil condenser for use with rotary evaporators for cooling with tap water in a vertical position. Condenser has a 45/40 lower inner drip tip joint, a 29/32 top outer joint, and removable hose connections using GL-14 glass threads. Condenser is supplied complete with plastic caps and serrated hose connections.
Catalog No. CG132910 Supplier Chemglass Life Sciences Supplier No. CG132910
Condenser, "B" Assembly, For Rotary Evaporator, Plastic Coated. Double coil condenser for use with rotary evaporators for cooling with tap water in a vertical position. Condenser has a 45/40 lower inner drip tip joint, a 29/32 top outer joint, and removable hose connections using GL-14 glass threads. Condenser is supplied complete with plastic caps and serrated hose connections.