Learn More
BUCHI Condensers for BUCHI Rotavapors
SDP
Catalog No. 01673000 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Rotavapor R-205, R-210/R-215
Rotavapor R-80
Material (Coating):
P+G
Plastic and Glass
Type:
Cold Trap Condenser
Condenser C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material (Coating) Type
01-673-000 Rotavapor R-80 Plastic and Glass Cold Trap Condenser
05-402-495 Rotavapor R-205, R-210/R-215 P+G Condenser C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-673-000 Supplier BUCHI Supplier No. 11075730
Add to Cart
Add to Cart
Original BUCHI glassware guarantees the highest levels of efficiency and safety thanks to high precision in manufacturing. For more than 80 years, BUCHI has been manufacturing innovative and durable glass parts to ease daily laboratory work.
  • High throughput due to advanced design of condensers
  • Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
  • Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
  • Maximum stability and safety with plastic and glass coating
Specifications
Material Glass
Type Cold Trap Condenser
For Use With (Application) Evaporator Condensation
For Use With (Equipment) Rotavapor R-80
Material (Coating) Plastic and Glass