Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Control Cable RJ 45 for Vacuum Pumps and Controllers
SDP

Catalog No. 05422875 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Vacuum pump and controllers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-422-875 Vacuum pump and controllers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-422-875 Supplier BUCHI Supplier No. 044989
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Communication cable. Connection between vacuum controller/interface and recirculating chiller or vacuum controller/interface and vacuum pump.

  • Gray Ethernet cable
  • 2m
  • Measuring range: 0-14000hPa (mbar)
  • Ambient temperature range: 10 to 40°C

Specifications

Color Gray
Type RJ45 Cat.5e Ethernet Cable
For Use With (Equipment) Vacuum pump and controllers
Length (Metric) 2 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.