Learn More
BUCHI Evaporating Flask Support Ring
SDP
Catalog No. 05403206 Shop All BUCHI Products
Change view
Click to view available options
Material:
Silicone
Catalog No. Material
05-403-206 Silicone
1 options
Catalog No. 05-403-206 Supplier BUCHI Supplier No. 048618
Add to Cart
Add to Cart
Flask holder for safely holding 50 to 5000mL evaporating and receiving flasks.
  • Flask ring made of non-sliding silicone
Specifications
Material Silicone
Type Flask Support
For Use With (Equipment) Evaporation system