Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Set of Bent Hose Barbs with GL14 threads for condensers
GSA_VASDP

Catalog No. 05422876 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
R-300
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-422-876 R-300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-422-876 Supplier BUCHI Supplier No. 037287
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains hose barbs, cap nuts, seals.

  • For use with Rotavapor
  • Hose barbs, set 4pcs, bent GL14, silicone seal

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

For Use With (Equipment) R-300
Type Hose Barb
Material Silicone
Plug Type Connection: GL14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.