Learn More
BUCHI Ball Joint Clamp for BUCHI condenser and receiving flask
SDP
Catalog No. 05422908 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
BUCHI Syncore Polyvap parallel evaporation systems
Catalog No. For Use With (Equipment)
05-422-908 BUCHI Syncore Polyvap parallel evaporation systems
1 options
Catalog No. 05-422-908 Supplier BUCHI Supplier No. 003275
Add to Cart
Add to Cart
Clamp is designed to work with ground glass ball joints on BUCHI condensers and receiving flasks.
  • Joint: SJ20/35
  • Fits all models of condensers
Specifications
Material Stainless Steel
Type Joint Clamp
For Use With (Equipment) BUCHI Syncore Polyvap parallel evaporation systems
Standard Taper Joint Number Center SJ35/20