Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments
SDP

Catalog No. p-4892050 Shop All BUCHI Products
Change view
Click to view available options
Product Type:
Calibration Kit
Calibration Set
Verification Kit
For Use With (Equipment):
BUCHI Melting Point Instrument
Melting Point Instruments
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
05-405-897 Calibration Kit Melting Point Instruments
10-987-120 Calibration Set BUCHI Melting Point Instrument
10-987-121 Verification Kit BUCHI Melting Point Instrument
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-405-897 Supplier BUCHI Supplier No. 037667
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.

Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.

  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C

Specifications

Product Type Calibration Kit
For Use With (Application) Instrument Calibration and Verification
For Use With (Equipment) Melting Point Instruments
Model B-540/545
Temperature Range (English) 50°F to 752°F
Temperature Range (Metric) 10°C to 400°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.