Learn More
BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments
SDP
Catalog No. 05404362 Shop All BUCHI Products
Change view
Click to view available options
Product Type:
Calibration Kit
Catalog No. Product Type
05-404-362 Calibration Kit
1 options
Catalog No. 05-404-362 Supplier BUCHI Supplier No. 037454
Add to Cart
Add to Cart
Includes
Vanillin, Caffeine, Phenacetin
Utilize the embedded firmware and software on your BUCHI melting point apparatus to calibrate and verify your instrument at any time. Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories.
Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.
  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C
Specifications
Product Type Calibration Kit
For Use With (Equipment) BUCHI B-540/545 Melting Point apparatus
Includes Vanillin, caffeine, phenacetin