Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Set of Bent and Straight Hose Barbs with GL14 threads for condensers
GSA_VASDP

Catalog No. 05422897 Shop All BUCHI Products
Change view
Click to view available options
Size:
GL14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
05-422-897 GL14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-422-897 Supplier BUCHI Supplier No. 038000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Package contains hose barbs, cap nuts, and seals for BUCHI Rotavapor™ vacuum and chiller connections.

  • Hose barb set, 6 pcs
  • Bent (4), straight (2)
  • GL14 caps
  • Silicone seals

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Description Hose Barb Set
Diameter (Metric) Outside 8 mm
For Use With Vacuum and chiller connections
Includes Bent (4), Straight (2)
Material Polypropylene
Overall Length (Metric) 30 mm
Size GL14
Sterility Non-sterile
Type Hose Barb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.