Learn More
BUCHI Valve Unit for Rotavapor RII
GSA_VASDP
Catalog No. 05412905 Shop All BUCHI Products
Change view
Click to view available options
Material:
Plastic, glass
Catalog No. Material
05-412-905 Plastic, glass
1 options
Catalog No. 05-412-905 Supplier BUCHI Supplier No. 047160
Add to Cart
Add to Cart
Valve unit includes Woulff bottle, 125mL, P+G, holder. Valve is meant to be used with a vacuum controller/interface and non-BUCHI vacuum source
  • Control house vacuum or 3rd party vacuum pump
  • Connects to BUCHI controllers for vacuum regulation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With Rotavapor RII (use only in combination with a vacuum controller/interface)
Includes Woulff bottle, 125mL; holder
Material Plastic, glass