Learn More
Chemglass Life Sciences Buchner Funnel, Table Top, Polyethylene, Perforated, 10.25in ID, Fixed Plate
SDP
Catalog No. CG141317
Encompass_Preferred
Buchner Funnel, Table Top, Polyethylene, Perforated, 10.25in ID, Fixed Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Welded hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Welded hose barbs allow the port to be as low as possible to the inside bottom of the funnel. Perforated filter plates are made of 4.8mm (3/16") thick HDPE with 4.8mm (3/16") perforations on 11mm (7/16") centers. Fixed plate is used for coarse filtration or use with a cloth or paper filter; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).
Catalog No. CG141317 Supplier Chemglass Life Sciences Supplier No. CG141317
Buchner Funnel, Table Top, Polyethylene, Perforated, 10.25in ID, Fixed Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Welded hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Welded hose barbs allow the port to be as low as possible to the inside bottom of the funnel. Perforated filter plates are made of 4.8mm (3/16") thick HDPE with 4.8mm (3/16") perforations on 11mm (7/16") centers. Fixed plate is used for coarse filtration or use with a cloth or paper filter; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).