Learn More
Chemglass Life Sciences Buchner Funnel, Table Top, Polyethylene, Perforated, 10.25in ID, Removable Plate
SDP
Catalog No. CG141318
Encompass_Preferred
Buchner Funnel, Table Top, Polyethylene, Perforated, 10.25in ID, Removable Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Welded hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Welded hose barbs allow the port to be as low as possible to the inside bottom of the funnel. Perforated filter plates are made of 4.8mm (3/16") thick HDPE with 4.8mm (3/16") perforations on 11mm (7/16") centers. Removable plate is for use in precious metals recovery; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).
Catalog No. CG141318 Supplier Chemglass Life Sciences Supplier No. CG141318
Buchner Funnel, Table Top, Polyethylene, Perforated, 10.25in ID, Removable Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Welded hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Welded hose barbs allow the port to be as low as possible to the inside bottom of the funnel. Perforated filter plates are made of 4.8mm (3/16") thick HDPE with 4.8mm (3/16") perforations on 11mm (7/16") centers. Removable plate is for use in precious metals recovery; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).