Learn More
Chemglass Life Sciences Buchner Funnel, Table Top, Polyethylene, Perforated, 24in ID, Fixed Plate
SDP
Catalog No. CG141321
Encompass_Preferred
Buchner Funnel, Table Top, Polyethylene, Perforated, 24in ID, Fixed Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Removable hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Perforated filter plates are made of 4.8mm (3/16") thick HDPE with 4.8mm (3/16") perforations on 11mm (7/16") centers. Fixed plates are for use with coarse filtration or use with a cloth or paper filter; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).
Catalog No. CG141321 Supplier Chemglass Life Sciences Supplier No. CG141321
Add to Cart
Add to Cart
Buchner Funnel, Table Top, Polyethylene, Perforated, 24in ID, Fixed Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Removable hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Perforated filter plates are made of 4.8mm (3/16") thick HDPE with 4.8mm (3/16") perforations on 11mm (7/16") centers. Fixed plates are for use with coarse filtration or use with a cloth or paper filter; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).