Learn More
Cole-Parmer Cole-Parmer Buchner Funnel, porcelain, 1150 mL, 1/ea
Catalog No. NC1375601 Shop All Cole-Parmer Products
Encompass_Preferred
Funnel includes fixed perforated plate and is glazed inside and out, except for the rim. Resistance to acids and alkalies is excellent, except for hydrofluoric acid. Autoclavable.
Catalog No. NC1375601 Supplier Cole-Parmer Supplier No. 1780015
Funnel includes fixed perforated plate and is glazed inside and out, except for the rim. Resistance to acids and alkalies is excellent, except for hydrofluoric acid. Autoclavable.