Learn More
Chemglass Life Sciences Buchner Funnel, 150mL, Perforated Plate
SDP
Catalog No. 501311031
Encompass_Preferred
Buchner Funnel, 150mL, Perforated Plate. Buchner funnels having a perforated plate rather than a fritted disc. Funnels accept filter paper size as listed.
Catalog No. 50-131-1031 Supplier Chemglass Life Sciences Supplier No. CG1402P02
Buchner Funnel, 150mL, Perforated Plate. Buchner funnels having a perforated plate rather than a fritted disc. Funnels accept filter paper size as listed.