Learn More
Chemglass Life Sciences Buchner Funnel, Table Top, Polyethylene, Porous, 18in ID, Coarse Porosity, Fixed Plate
SDP
Catalog No. CG141306
Encompass_Preferred
Buchner Funnel, Table Top, Polyethylene, Porous, 18in ID, Coarse Porosity, Fixed Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Removable hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Filter plates are supported by a multiple ring grid; HDPE, 6.4mm (1/4") thick, with a non-porous ring at the periphery of the plate which seals filter paper when used with porous filter plates. Coarse porosity is 90-130 microns; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).
Catalog No. CG141306 Supplier Chemglass Life Sciences Supplier No. CG141306
Add to Cart
Add to Cart
Buchner Funnel, Table Top, Polyethylene, Porous, 18in ID, Coarse Porosity, Fixed Plate. Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate. Removable hose barb connector accepts 12.7mm (1/2") I.D. tube and threads into 3/4" NPT bung on the funnel. Filter plates are supported by a multiple ring grid; HDPE, 6.4mm (1/4") thick, with a non-porous ring at the periphery of the plate which seals filter paper when used with porous filter plates. Coarse porosity is 90-130 microns; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF).