Learn More
TARGETMOL CHEMICALS INC BUCINDOLOL FORMATE 5MG
SDP
Catalog No. 502581700
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bucindolol Formatel is a blocker of (beta)1-adrenergic receptor. Bucindolol exhibits intrinsic sympathomimetic activity. Bucindolol can be used in the research of heart failure.
Catalog No. 50-258-1700 Supplier TARGETMOL CHEMICALS INC Supplier No. T10631L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bucindolol Formatel is a blocker of (beta)1-adrenergic receptor. Bucindolol exhibits intrinsic sympathomimetic activity. Bucindolol can be used in the research of heart failure.