Learn More
Med Vet International Miltex Vantage Buck Ear Curette, 6.75in., Straight, Size 00 Tip
Catalog No. 502830286
Encompass_Preferred
The Miltex Vantage V919-280 Buck ear curette is made from high quality Japanese stainless steel and measures 6.75" in length. It is single-ended and has a straight tip, size 00.
Catalog No. 50-283-0286 Supplier Med Vet International Supplier No. MILV919280
The Miltex Vantage V919-280 Buck ear curette is made from high quality Japanese stainless steel and measures 6.75" in length. It is single-ended and has a straight tip, size 00.