Learn More
MSC Arborjet AccuFlo Drench Bucket, 4 cu in, Polyethylene, 1 ea.
Catalog No. 502818070
Encompass
The AccuFlo Drench Kit Bucket Assembly is a new product designed for applying fluid nutrients, pesticides, and fungicides in lawn, garden, and tree care applications. It provides consistent, repeatable results.
  • Backpack sprayer type
  • Polyethylene tank material
  • Four cubic inch volume capacity
  • Cone, flat, and stream spray patterns
  • Chemical safe
  • In-tank filter
  • Aluminum and brass wand material
  • Brass nozzle material
  • Synthetic rubber seal/gasket material
  • Includes battery
  • Six inch opening diameter
  • Kit includes AccuFlo device, USB charging cable, training manual, 4 L measuring pitcher, 500 mL measuring pitcher, handheld pick axe
Catalog No. 50-281-8070 Supplier MSC Supplier No. 11669108
The AccuFlo Drench Kit Bucket Assembly is a new product designed for applying fluid nutrients, pesticides, and fungicides in lawn, garden, and tree care applications. It provides consistent, repeatable results.
  • Backpack sprayer type
  • Polyethylene tank material
  • Four cubic inch volume capacity
  • Cone, flat, and stream spray patterns
  • Chemical safe
  • In-tank filter
  • Aluminum and brass wand material
  • Brass nozzle material
  • Synthetic rubber seal/gasket material
  • Includes battery
  • Six inch opening diameter
  • Kit includes AccuFlo device, USB charging cable, training manual, 4 L measuring pitcher, 500 mL measuring pitcher, handheld pick axe