Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Bucket Lid for 12.5 qt. Utility Bucket
SDP

Catalog No. 22444928 Shop All Mopec Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-444-928 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-928 Supplier Mopec Supplier No. BA006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bucket lid can be used for a variety of facility needs.

  • Made of non-corrosive stainless steel

Specifications

Product Type Lid
Graduations No
Material Stainless Steel
Color Silver
For Use With (Equipment) 12.5 qt. Bucket
Handle Material Stainless Steel
Includes Lid
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.