Learn More
Beckman Coulter BUCKET ORING, SET OF 8, 368455
Catalog No. NC2011139
Encompass
Set of eight replacement O-rings for the 500 mL buckets of the TS-5.1-500 swinging bucket rotor
Catalog No. NC2011139 Supplier Beckman Coulter Supplier No. 368455
Set of eight replacement O-rings for the 500 mL buckets of the TS-5.1-500 swinging bucket rotor