Learn More
Beckman Coulter Beckman Coulter Bucket set for SW 41 Ti rotor, set of 6 (replacement buckets with caps and gaskets)
Catalog No. NC2876184
Encompass
Replacement bucket set for swinging-bucket ultracentrifuge rotors. This set contains six titanium buckets and includes matching caps and Buna N rubber gaskets for each bucket, supplied as replacement or spare parts to maintain sealing integrity and rotor performance.
  • Contains six titanium buckets with durable finish.
  • Includes caps and Buna N rubber gaskets for each bucket.
  • Caps are anodized aluminum for corrosion resistance.
  • Gaskets maintain an airtight seal; keep lightly coated with silicone vacuum grease.
  • Gaskets should be inspected regularly and replaced when worn, typically every six months.
  • Intended for use as replacement or spare parts to support routine rotor maintenance.
Catalog No. NC2876184 Supplier Beckman Coulter Supplier No. 333790
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Replacement bucket set for swinging-bucket ultracentrifuge rotors. This set contains six titanium buckets and includes matching caps and Buna N rubber gaskets for each bucket, supplied as replacement or spare parts to maintain sealing integrity and rotor performance.
  • Contains six titanium buckets with durable finish.
  • Includes caps and Buna N rubber gaskets for each bucket.
  • Caps are anodized aluminum for corrosion resistance.
  • Gaskets maintain an airtight seal; keep lightly coated with silicone vacuum grease.
  • Gaskets should be inspected regularly and replaced when worn, typically every six months.
  • Intended for use as replacement or spare parts to support routine rotor maintenance.