Learn More
Micronova™ BucketBinders™ Bucket Connectors
BIOPHARMASDP
Catalog No. 19263628 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
19-263-628 Stainless Steel
1 options
This item is not returnable. View return policy
Catalog No. 19-263-628 Supplier Micronova™ Supplier No. BB01
Add to Cart
Add to Cart
This item is not returnable. View return policy
Bucket connecting hardware for BucketBinder™ Multi Bucket System
The BucketBinder™ kit includes 2 or 3 polypropylene buckets with casters and one or two sets of BucketBinders (bucket connectors). The system offers easier cleaning and storage - in both small and large controlled environments and the sterile core.
Specifications
Product Type Bucket Connector, Hardware
Material Stainless Steel
Autoclavable Autoclavable
For Use With (Application) Cleanroom Surface Cleaning and Disinfecting
For Use With (Equipment) B-7AC Rectangular Bucket
Height (Metric) 25 cm
Includes Bucket Connector
Quantity 1 Pair
Sterility Non-sterile