Learn More
Thermo Scientific™ Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
GSA_VA
Catalog No. 75005721 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Sealed Bucket
Unsealed Bucket
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
75-005-721 Sealed Bucket
75-005-723 Unsealed Bucket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-005-721 Supplier Thermo Scientific™ Supplier No. 75005721
  • $2,680.00
    $2,185.65 / Each of 1
    Save $494.35
    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Max Tube Dimensions: Height <33 mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) M10 rotor
Capacity 4 Standard or 2 Midi-Deepwell plates
Product Type Sealed Bucket
Description Sealed Bucket; Capacity: 4 Standard or 2 Midi-Deepwell plates (Set of 2)