Learn More
Medchemexpress LLC Bucladesine (sodium) | 16980-89-5 | 99.4% | 1 ML
SDP
Catalog No. 5000409275
Encompass_Preferred
Bucladesine sodium is a stabilized cyclic AMP (cAMP) analog that acts as a selective PKA activator. It increases intracellular cAMP levels and also functions as a phosphodiesterase (PDE) inhibitor. This compound possesses anti-inflammatory activity and has been utilized for impaired wound healing research.
  • Acts as a selective PKA activator
  • Increases intracellular cAMP levels
  • Functions as a phosphodiesterase (PDE) inhibitor
  • Exhibits anti-inflammatory activity
  • Can be used for impaired wound healing research
  • Increases choline acetyltransferase (ChAT) and vesicular acetylcholine transporter (VAChT) mRNA levels in PC12 cells
  • Improves spatial memory retention in vivo studies
  • Supplied as a white to off-white solid
  • Soluble in water and DMSO
Catalog No. 50-004-09275 Supplier Medchemexpress LLC Supplier No. HYB076410MM/1ML
Bucladesine sodium is a stabilized cyclic AMP (cAMP) analog that acts as a selective PKA activator. It increases intracellular cAMP levels and also functions as a phosphodiesterase (PDE) inhibitor. This compound possesses anti-inflammatory activity and has been utilized for impaired wound healing research.
  • Acts as a selective PKA activator
  • Increases intracellular cAMP levels
  • Functions as a phosphodiesterase (PDE) inhibitor
  • Exhibits anti-inflammatory activity
  • Can be used for impaired wound healing research
  • Increases choline acetyltransferase (ChAT) and vesicular acetylcholine transporter (VAChT) mRNA levels in PC12 cells
  • Improves spatial memory retention in vivo studies
  • Supplied as a white to off-white solid
  • Soluble in water and DMSO