Learn More
TARGETMOL CHEMICALS INC Bucladesine sodium 200MG
SDP
Catalog No. 502876429
Encompass_Preferred
Also available in 2 g, 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Bucladesine sodium (DC2797) is a cAMP analog with cell-permeable properties. Bucladesine sodium is also a cAMP-dependent protein kinase (PKA) activator and a phosphodiester (PDE) inhibitor. Bucladesine sodium has anti-inflammatory activity. Purity 98.34%
Catalog No. 50-287-6429 Supplier TARGETMOL CHEMICALS INC Supplier No. T1418200MG
Also available in 2 g, 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Bucladesine sodium (DC2797) is a cAMP analog with cell-permeable properties. Bucladesine sodium is also a cAMP-dependent protein kinase (PKA) activator and a phosphodiester (PDE) inhibitor. Bucladesine sodium has anti-inflammatory activity. Purity 98.34%