Learn More
Cayman Chemical Bucladesine (sodium salt), Purity: ≥95%, CAS: 16980-89-5, 100 mg
SDP
Catalog No. 502971142
Encompass_Preferred
A cell-permeable, cyclic nucleotide derivative that mimics the action of endogenous cAMP and is a phosphodiesterase inhibitor
Catalog No. 50-297-1142 Supplier Cayman Chemical Supplier No. 14408100 MG
A cell-permeable, cyclic nucleotide derivative that mimics the action of endogenous cAMP and is a phosphodiesterase inhibitor