Learn More
Cayman Chemical Bucladesine (sodium salt), Purity: ≥95%, CAS: 16980-89-5, 50 mg
SDP
Catalog No. 502972116
Encompass_Preferred
A cell-permeable, cyclic nucleotide derivative that mimics the action of endogenous cAMP and is a phosphodiesterase inhibitor
Catalog No. 50-297-2116 Supplier Cayman Chemical Supplier No. 1440850 MG
A cell-permeable, cyclic nucleotide derivative that mimics the action of endogenous cAMP and is a phosphodiesterase inhibitor