Learn More
Cayman Chemical Buclizine (hydrochloride), Purity: ≥98%, CAS: 129-74-8, 100 mg
SDP
Catalog No. 502934287
Encompass_Preferred
An antihistamine; reduces bronchoconstriction and lethality induced by aerosolized histamine in guinea pigs at 1 mg/kg; induces gross malformations and bone malformations in rats from 40-200 mg/kg; inhibits cancer cell growth by binding to TCTP and inducing cell differentiation
Catalog No. 50-293-4287 Supplier Cayman Chemical Supplier No. 25641100 MG
An antihistamine; reduces bronchoconstriction and lethality induced by aerosolized histamine in guinea pigs at 1 mg/kg; induces gross malformations and bone malformations in rats from 40-200 mg/kg; inhibits cancer cell growth by binding to TCTP and inducing cell differentiation