Learn More
Cayman Chemical BuclIzInhydrochlorIde 50mg
SDP
Catalog No. 502936743
Encompass_Preferred
An antihistamine; reduces bronchoconstriction and lethality induced by aerosolized histamine in guinea pigs at 1 mg/kg; induces gross malformations and bone malformations in rats from 40-200 mg/kg; inhibits cancer cell growth by binding to TCTP and inducing cell differentiation
Catalog No. 50-293-6743 Supplier Cayman Chemical Supplier No. 2564150 MG
An antihistamine; reduces bronchoconstriction and lethality induced by aerosolized histamine in guinea pigs at 1 mg/kg; induces gross malformations and bone malformations in rats from 40-200 mg/kg; inhibits cancer cell growth by binding to TCTP and inducing cell differentiation