Learn More
Aviva Systems Biology Corp BUD31 Antibody - middle region (ARP38402_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38402_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BUD31. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BUD31. Alternate names: G10, EDG2, EDG-2, fSAP17, YCR063WApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-56640