Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Buddlejasaponin IVb | 152580-79-5 | 5 MG
SDP

Catalog No. 5000421899
Encompass_Preferred

Buddlejasaponin IVb is a triterpene saponin isolated from Clinopodium chinense (Benth.) O. Kuntze. It exhibits hemostasis efficacy and has been shown to shorten thrombin time (TT) by 20.6%. This product is intended for research use only.

  • Isolated from Clinopodium chinense (Benth.) O. Kuntze
  • Exhibits hemostasis efficacy
  • Shortens thrombin time (TT) by 20.6%
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-21899 Supplier Medchemexpress LLC Supplier No. HYN21385MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Buddlejasaponin IVb is a triterpene saponin isolated from Clinopodium chinense (Benth.) O. Kuntze. It exhibits hemostasis efficacy and has been shown to shorten thrombin time (TT) by 20.6%. This product is intended for research use only.

  • Isolated from Clinopodium chinense (Benth.) O. Kuntze
  • Exhibits hemostasis efficacy
  • Shortens thrombin time (TT) by 20.6%
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.