Learn More
Medchemexpress LLC Budesonide-d8 | 1105542-94-6 | MFCD07369194 | 97.6% | 438.58 g/mol | C25H26D8O6 | 5 MG
SDP
Catalog No. 5000222354
Encompass_Preferred
Budesonide-d8 is the deuterium-labeled analog of budesonide intended for use as an internal standard and reference material in bioanalytical and pharmacokinetic studies. Its mass-shifted isotopic composition allows specific detection and accurate quantitation by mass spectrometry while closely matching the chemical behavior of the unlabeled analyte.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • Enables accurate quantitation and reliable peak identification.
  • Suitable for pharmacokinetic, metabolic, and bioanalytical studies.
  • High purity for analytical reproducibility (approximately 97.6%).
  • Provided in small-mass formats for method development and validation.
Catalog No. 50-002-22354 Supplier Medchemexpress LLC Supplier No. HY13580S5MG
Budesonide-d8 is the deuterium-labeled analog of budesonide intended for use as an internal standard and reference material in bioanalytical and pharmacokinetic studies. Its mass-shifted isotopic composition allows specific detection and accurate quantitation by mass spectrometry while closely matching the chemical behavior of the unlabeled analyte.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • Enables accurate quantitation and reliable peak identification.
  • Suitable for pharmacokinetic, metabolic, and bioanalytical studies.
  • High purity for analytical reproducibility (approximately 97.6%).
  • Provided in small-mass formats for method development and validation.