Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Budesonide impurity C | 1040085-99-1 | MFCD29920082 | 98.6% | 430.53 g/mol | C25H34O6 | 10 MG
SDP

Catalog No. 5000215640
Encompass_Preferred

Budesonide impurity C is a chemical impurity of the corticosteroid budesonide, supplied as a research reference material for analytical characterization, impurity profiling, and analytical method development in laboratory and quality-control workflows.

  • Used for impurity profiling and analytical method development.
  • Purity 98.6%.
  • Molecular formula C25H34O6.
  • Molecular weight 430.53 g/mol.
  • CAS number 1040085-99-1 for unambiguous identification.
  • Available in small research pack sizes (10 mg).

Catalog No. 50-002-15640 Supplier Medchemexpress LLC Supplier No. HY10008710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Budesonide impurity C is a chemical impurity of the corticosteroid budesonide, supplied as a research reference material for analytical characterization, impurity profiling, and analytical method development in laboratory and quality-control workflows.

  • Used for impurity profiling and analytical method development.
  • Purity 98.6%.
  • Molecular formula C25H34O6.
  • Molecular weight 430.53 g/mol.
  • CAS number 1040085-99-1 for unambiguous identification.
  • Available in small research pack sizes (10 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.