Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Budesonide 98.0+%, TCI America™
SDP

Catalog No. B39091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39091G 1 g
B3909200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39091G Supplier TCI America Supplier No. B39091G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51333-22-3
Molecular Formula C25H34O6
Molecular Weight (g/mol) 430.54
MDL Number MFCD00083259,MFCD00138154
InChI Key VOVIALXJUBGFJZ-VXKMTNQYSA-N
Synonym (+)-16alpha,17alpha-Butylidenedioxy-11beta,21-dihydroxy-1,4-pregnadiene-3,20-dione
PubChem CID 73759663
IUPAC Name (1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
SMILES CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 430.54
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Budesonide
RTECSNumber TU3723000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.