Learn More
Anton Paar Budget-friendly with best build-quality, precisionand temperature control with Peltier System.Up to 10 methods: Refractive index and Brix preset,plus 8 optional methodsHigh accuracy measurements and easy label printing.
Catalog No. 5000580522
Encompass_Preferred
Measuring specifications Abbemat 3101Refractive indexRange: 1.30 nD to 1.66 nDMax. resolution: 0.00001 nDAccuracy: ±0.00004 nDRepeatability: ±0.000015 nDBrixRange: 0 °Brix to 100 °BrixDisplay resolution: 0.001 °BrixAccuracy: ±0.03 °BrixTemperatureTemperature range: 10 °C to 70 °CTemperature probe accuracy: ±0.03 °CTemperature probe stability: ±0.002 °CDisplay7" Hybrid touchCommunication interfaces: USB (printer)Data storage: Label printerGeneralMin. sample volume 70 uLMax. allowed pressure in micro flow cell: 10 barAmbient temperature: 5 °C to 40 °CMaterials in contact with the samplePrism: Synthetic sapphireSample well: Stainless steel, optionally Ni-alloySeal: FFKM (perfluoroelastomer)CompoundsLight source: LED light source, average lifetime > 100,000 hoursWavelength: 589.3 nm (by wavelength-adjusted interference filter)Power requirements: 100 VAC to 240 VAC +10 %/-15 %, 50/60 Hz, min.DimensionsW x H x D 225 mm x 115 mm x 330 mmWeight 3.5 kg
Catalog No. 50-005-80522 Supplier Anton Paar Supplier No. 313363
Measuring specifications Abbemat 3101Refractive indexRange: 1.30 nD to 1.66 nDMax. resolution: 0.00001 nDAccuracy: ±0.00004 nDRepeatability: ±0.000015 nDBrixRange: 0 °Brix to 100 °BrixDisplay resolution: 0.001 °BrixAccuracy: ±0.03 °BrixTemperatureTemperature range: 10 °C to 70 °CTemperature probe accuracy: ±0.03 °CTemperature probe stability: ±0.002 °CDisplay7" Hybrid touchCommunication interfaces: USB (printer)Data storage: Label printerGeneralMin. sample volume 70 uLMax. allowed pressure in micro flow cell: 10 barAmbient temperature: 5 °C to 40 °CMaterials in contact with the samplePrism: Synthetic sapphireSample well: Stainless steel, optionally Ni-alloySeal: FFKM (perfluoroelastomer)CompoundsLight source: LED light source, average lifetime > 100,000 hoursWavelength: 589.3 nm (by wavelength-adjusted interference filter)Power requirements: 100 VAC to 240 VAC +10 %/-15 %, 50/60 Hz, min.DimensionsW x H x D 225 mm x 115 mm x 330 mmWeight 3.5 kg