Learn More
Research Products International Corp Budget-Solve, 1 CS
SDP
Catalog No. 501977072
Encompass_Preferred
Budget-Solve is an economical ready-to-use cocktail for liquid scintillation counting. No dilution is required. It contains a blend of quench resistant fluors in an efficient emulsifier system. Budget-Solve is an economical product of exceptional performance. It is designed to handle a wide range of liquid scintillation counting samples.
WARNING! This product can expose you to Methyl alcohol, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7072 Supplier Research Products International Corp Supplier No. 111167CS
Budget-Solve is an economical ready-to-use cocktail for liquid scintillation counting. No dilution is required. It contains a blend of quench resistant fluors in an efficient emulsifier system. Budget-Solve is an economical product of exceptional performance. It is designed to handle a wide range of liquid scintillation counting samples.
WARNING! This product can expose you to Methyl alcohol, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.