Learn More
Medchemexpress LLC Budiodarone tartrate | 478941-93-4 | 98.1% | 853.43 | 1 ML
SDP
Catalog No. 5000362994
Encompass_Preferred
Budiodarone tartrate (ATI-2042 tartrate) is a chemical analogue of Amiodarone that exhibits balanced inhibitory activity across multiple cardiac ion channels, specifically potassium, sodium, and calcium channels. It functions as an antiarrhythmic agent.
  • Chemical analogue of Amiodarone
  • Exhibits balanced inhibitory activity across multiple cardiac ion channels
  • Inhibits potassium channels
  • Inhibits sodium channels
  • Inhibits calcium channels
  • Functions as an antiarrhythmic agent
Catalog No. 50-003-62994 Supplier Medchemexpress LLC Supplier No. HY14834A10MM/1ML
Budiodarone tartrate (ATI-2042 tartrate) is a chemical analogue of Amiodarone that exhibits balanced inhibitory activity across multiple cardiac ion channels, specifically potassium, sodium, and calcium channels. It functions as an antiarrhythmic agent.
  • Chemical analogue of Amiodarone
  • Exhibits balanced inhibitory activity across multiple cardiac ion channels
  • Inhibits potassium channels
  • Inhibits sodium channels
  • Inhibits calcium channels
  • Functions as an antiarrhythmic agent