Learn More
Medchemexpress LLC Budiodarone tartrate | 478941-93-4 | 98.1% | 853.43 | 100 MG
SDP
Catalog No. 5000362816
Encompass_Preferred
Budiodarone tartrate is a chemical analogue of Amiodarone, exhibiting balanced inhibitory activity across multiple cardiac ion channels, including potassium, sodium, and calcium channels. It functions as an antiarrhythmic agent. The compound is characterized by a short plasma half-life of 7 hours and a lower volume of distribution at 13 L/kg.
  • Functions as a chemical analogue of Amiodarone.
  • Exhibits balanced, multiple cardiac ion channel inhibiting activity, targeting potassium, sodium, and calcium channels.
  • Acts as an antiarrhythmic agent.
  • Demonstrates a short plasma half-life of 7 hours.
  • Possesses a lower volume of distribution of 13 L/kg.
Catalog No. 50-003-62816 Supplier Medchemexpress LLC Supplier No. HY14834A100MG
Budiodarone tartrate is a chemical analogue of Amiodarone, exhibiting balanced inhibitory activity across multiple cardiac ion channels, including potassium, sodium, and calcium channels. It functions as an antiarrhythmic agent. The compound is characterized by a short plasma half-life of 7 hours and a lower volume of distribution at 13 L/kg.
  • Functions as a chemical analogue of Amiodarone.
  • Exhibits balanced, multiple cardiac ion channel inhibiting activity, targeting potassium, sodium, and calcium channels.
  • Acts as an antiarrhythmic agent.
  • Demonstrates a short plasma half-life of 7 hours.
  • Possesses a lower volume of distribution of 13 L/kg.