Learn More
Medchemexpress LLC Budiodarone tartrate | 478941-93-4 | 98.14% | 853.43 | 25 MG
SDP
Catalog No. 5000363568
Encompass_Preferred
Budiodarone tartrate (ATI-2042) is a chemical analogue of Amiodarone that acts as an antiarrhythmic agent. It inhibits multiple cardiac ion channels, including potassium, sodium, and calcium channels.
  • Acts as an antiarrhythmic agent
  • Inhibits multiple cardiac ion channels
  • Exhibits balanced potassium, sodium, and calcium channel inhibition
  • Chemical analogue of Amiodarone
  • High purity of 98.14%
Catalog No. 50-003-63568 Supplier Medchemexpress LLC Supplier No. 25MGHY14834A
Budiodarone tartrate (ATI-2042) is a chemical analogue of Amiodarone that acts as an antiarrhythmic agent. It inhibits multiple cardiac ion channels, including potassium, sodium, and calcium channels.
  • Acts as an antiarrhythmic agent
  • Inhibits multiple cardiac ion channels
  • Exhibits balanced potassium, sodium, and calcium channel inhibition
  • Chemical analogue of Amiodarone
  • High purity of 98.14%