Learn More
Medchemexpress LLC Budiodarone tartrate | 478941-93-4 | 98.14% | 853.43 | 5 MG
SDP
Catalog No. 5000362877
Encompass_Preferred
Budiodarone tartrate is a chemical analogue of Amiodarone with balanced, multiple cardiac ion channel (potassium, sodium and calcium channels) inhibiting activity. It acts as an antiarrhythmic agent.
  • Chemical analogue of Amiodarone.
  • Inhibits multiple cardiac ion channels (potassium, sodium, and calcium channels).
  • Antiarrhythmic agent.
  • Has a short plasma half-life (7 h) and a lower volume of distribution (13 L/kg) in vitro.
Catalog No. 50-003-62877 Supplier Medchemexpress LLC Supplier No. HY14834A5MG
Budiodarone tartrate is a chemical analogue of Amiodarone with balanced, multiple cardiac ion channel (potassium, sodium and calcium channels) inhibiting activity. It acts as an antiarrhythmic agent.
  • Chemical analogue of Amiodarone.
  • Inhibits multiple cardiac ion channels (potassium, sodium, and calcium channels).
  • Antiarrhythmic agent.
  • Has a short plasma half-life (7 h) and a lower volume of distribution (13 L/kg) in vitro.