Learn More
Medchemexpress LLC Budiodarone tartrate | 478941-93-4 | 98.14% | 853.43 | 50 MG
SDP
Catalog No. 5000361472
Encompass_Preferred
Budiodarone tartrate, also known as ATI-2042 tartrate, is a chemical analogue of Amiodarone. This antiarrhythmic agent exhibits balanced and multiple cardiac ion channel inhibiting activity.
  • Inhibits potassium, sodium, and calcium cardiac ion channels
  • Possesses a short plasma half-life (7 h)
  • Has a lower volume of distribution (13 L/kg)
Catalog No. 50-003-61472 Supplier Medchemexpress LLC Supplier No. HY14834A50MG
Budiodarone tartrate, also known as ATI-2042 tartrate, is a chemical analogue of Amiodarone. This antiarrhythmic agent exhibits balanced and multiple cardiac ion channel inhibiting activity.
  • Inhibits potassium, sodium, and calcium cardiac ion channels
  • Possesses a short plasma half-life (7 h)
  • Has a lower volume of distribution (13 L/kg)