Learn More
Medchemexpress LLC Budipine | 57982-78-2 | 293.45 | 1 ML
SDP
Catalog No. 5000446084
Encompass_Preferred
Budipine is an anti-parkinson agent that acts as a substrate of P-glycoprotein (P-gp) and an N-methyl-d-aspartate (NMDA) antagonist. It exhibits indirect dopaminergic effects by improving dopamine release and inhibiting monoamine oxidase type B (MAO-B). This product is suitable for research related to CNS disorders, including Parkinson's disease.
  • Functions as an anti-parkinson agent
  • Acts as a substrate of P-glycoprotein (P-gp)
  • Functions as an N-methyl-d-aspartate (NMDA) antagonist
  • Exhibits indirect dopaminergic effects
  • Inhibits monoamine oxidase type B (MAO-B)
  • Useful for research on CNS disorders, including Parkinson's disease
Catalog No. 50-004-46084 Supplier Medchemexpress LLC Supplier No. HYW00160110MM/1ML
Budipine is an anti-parkinson agent that acts as a substrate of P-glycoprotein (P-gp) and an N-methyl-d-aspartate (NMDA) antagonist. It exhibits indirect dopaminergic effects by improving dopamine release and inhibiting monoamine oxidase type B (MAO-B). This product is suitable for research related to CNS disorders, including Parkinson's disease.
  • Functions as an anti-parkinson agent
  • Acts as a substrate of P-glycoprotein (P-gp)
  • Functions as an N-methyl-d-aspartate (NMDA) antagonist
  • Exhibits indirect dopaminergic effects
  • Inhibits monoamine oxidase type B (MAO-B)
  • Useful for research on CNS disorders, including Parkinson's disease