Learn More
Medchemexpress LLC Budipine | 57982-78-2 | 293.45 | 5 MG
SDP
Catalog No. 5000447002
Encompass_Preferred
Budipine is an anti-parkinson agent that functions as a substrate of P-glycoprotein (P-gp), mediating its uptake into the brain. It also acts as an N-methyl-d-aspartate (NMDA) antagonist and exhibits indirect dopaminergic effects by improving dopamine release and inhibiting monoamine oxidase type B (MAO-B). Budipine is suitable for research on central nervous system (CNS) disorders, including Parkinson's disease.
  • P-glycoprotein (P-gp) substrate
  • N-methyl-d-aspartate (NMDA) antagonist
  • Improves dopamine release
  • Inhibits monoamine oxidase type B (MAO-B)
  • Used in research for CNS disorders, including Parkinson's disease
Catalog No. 50-004-47002 Supplier Medchemexpress LLC Supplier No. HYW0016015MG
Budipine is an anti-parkinson agent that functions as a substrate of P-glycoprotein (P-gp), mediating its uptake into the brain. It also acts as an N-methyl-d-aspartate (NMDA) antagonist and exhibits indirect dopaminergic effects by improving dopamine release and inhibiting monoamine oxidase type B (MAO-B). Budipine is suitable for research on central nervous system (CNS) disorders, including Parkinson's disease.
  • P-glycoprotein (P-gp) substrate
  • N-methyl-d-aspartate (NMDA) antagonist
  • Improves dopamine release
  • Inhibits monoamine oxidase type B (MAO-B)
  • Used in research for CNS disorders, including Parkinson's disease