Learn More
AdipoGen Bufalin (10 mg)
SDP
Supplier:  AdipoGen AGCN20533M010
Encompass_Preferred
Chemical. CAS: 465-21-4. Formula: C24H34O4. MW: 386.5. Isolated from venenum bufonis. Bufalin is a cardiotonic steroid with anaesthetic, blood pressure stimulatory and respiratory activities. It is a potent Na+/K+-ATPase transporter inhibitor (Kd = 42, 45, and 40nM for the alpha1, alpha2, and alpha3 subunits, respectively). Bufalin has anticancer effects in a variety of cancer cell lines both in vitro and in vivo. It inhibits cancer cell proliferation, induces cell cycle arrest, induces apoptosis and autophagy, inhibits angiogenesis and neovascularization, induces cell differentiation, inhibits cancer metastasis and invasion, and enhances chemotherapeutic drug sensitivity through multiple kinase targets and also the Na+/K+-ATPase transporter.
Catalog No. 50-205-3714