Learn More
Apexbio Technology LLC Bufexamac 2438-72-4 50mg
SDP
Catalog No. 502240660
Encompass_Preferred
Bufexamac (CAS 2438-72-4) is a small molecule that functions as an inhibitor of cyclooxygenase (COX) modulating enzymatic pathways involved in inflammatory responses It has been shown to promote the release of interferon-alpha (IFN- ) with an EC50 of 8 9 M indicating its potency in modulating cytokine production Bufexamac is utilized in biomedical research to investigate COX-mediated signaling pathways and the regulation of interferon-related immune responses
This item is not returnable. View return policy
Catalog No. 50-224-0660 Supplier Apexbio Technology LLC Supplier No. B144350
Bufexamac (CAS 2438-72-4) is a small molecule that functions as an inhibitor of cyclooxygenase (COX) modulating enzymatic pathways involved in inflammatory responses It has been shown to promote the release of interferon-alpha (IFN- ) with an EC50 of 8 9 M indicating its potency in modulating cytokine production Bufexamac is utilized in biomedical research to investigate COX-mediated signaling pathways and the regulation of interferon-related immune responses