Learn More
NEOGEN Buffered Listeria Enrichment Broth, 10 kg
SDP
Catalog No. 502015302
Encompass_Preferred
Buffered Listeria Enrichment Broth is used for selective enrichment of Listeria spp and is not intended for use in the diagnosis of disease or other conditions in humans.Listeria monocytogenes, described first in 1926 by Murray, Webb, and Swann, is an extensive problem in public health and food industries. Evidence from outbreaks of listeriosis has indicated that the principle route of transmission is via consumption of foodstuffs contaminated with Listeria monocytogenes. Implicated vehicles of transmission include turkey, frankfurters, coleslaw, pasteurized milk, Mexican style cheese, and pate. Listeria spp. are ubiquitous in nature, being present in a wide range of unprocessed foods as well as in soil, sewage, and river water.For more information visit https://www.neogen.com/solutions/Microbiology/buffered-listeria-enrichment-broth
Catalog No. 50-201-5302 Supplier NEOGEN Supplier No. 700003103
Buffered Listeria Enrichment Broth is used for selective enrichment of Listeria spp and is not intended for use in the diagnosis of disease or other conditions in humans.Listeria monocytogenes, described first in 1926 by Murray, Webb, and Swann, is an extensive problem in public health and food industries. Evidence from outbreaks of listeriosis has indicated that the principle route of transmission is via consumption of foodstuffs contaminated with Listeria monocytogenes. Implicated vehicles of transmission include turkey, frankfurters, coleslaw, pasteurized milk, Mexican style cheese, and pate. Listeria spp. are ubiquitous in nature, being present in a wide range of unprocessed foods as well as in soil, sewage, and river water.For more information visit https://www.neogen.com/solutions/Microbiology/buffered-listeria-enrichment-broth