Learn More
NEOGEN Buffered Sodium Chloride - Peptone Broth pH 7.0, 500 g
SDP
Catalog No. 502015603
Encompass_Preferred
Buffered Sodium Chloride-Peptone Broth pH 7.0 is the diluent specified by the Harmonized Pharmacopoeia for the microbiological examination of non-sterile products andconforms to Harmonized USP/EP/JP Requirements in a laboratory setting. Buffered Sodium Chloride-Peptone Broth pH 7.0 is not intended for use in the diagnosis of disease or other conditions in humans.Buffered Sodium Chloride-Peptone Broth pH 7.0 is recommended by the Harmonized Pharmacopoeia (USP/EP/JP) for the microbiological examination of non-sterile products. The low level of peptone lessens the physiological shock experienced by microorganism when suspended in a diluent. The dual phosphate components create a buffered environment and sodium chloride maintains the osmotic balance.For more information visit https://www.neogen.com/solutions/Microbiology/buffered-sodium-chloride-peptone-broth-ph-7
Catalog No. 50-201-5603 Supplier NEOGEN Supplier No. 700003420
Buffered Sodium Chloride-Peptone Broth pH 7.0 is the diluent specified by the Harmonized Pharmacopoeia for the microbiological examination of non-sterile products andconforms to Harmonized USP/EP/JP Requirements in a laboratory setting. Buffered Sodium Chloride-Peptone Broth pH 7.0 is not intended for use in the diagnosis of disease or other conditions in humans.Buffered Sodium Chloride-Peptone Broth pH 7.0 is recommended by the Harmonized Pharmacopoeia (USP/EP/JP) for the microbiological examination of non-sterile products. The low level of peptone lessens the physiological shock experienced by microorganism when suspended in a diluent. The dual phosphate components create a buffered environment and sodium chloride maintains the osmotic balance.For more information visit https://www.neogen.com/solutions/Microbiology/buffered-sodium-chloride-peptone-broth-ph-7