Learn More
Surmodics Inc StabilCoat™ Immunoassay Stabilizer, 1L
SDP
Catalog No. 50949938
Encompass_Preferred
StabilCoat™ Immunoassay Stabilizer, and the 5X concentrate, effectively preserves the conformation and activity of dried proteins while simultaneously blocking the surface to reduce non-specific binding. The product can be used to stabilize antibodies, antigens or enzymes on an assortment of immunoassay components including polystyrene plates, tubes, glass, membranes, and filter paper.
Catalog No. 50-949-938 Supplier Surmodics Inc Supplier No. SC011000
StabilCoat™ Immunoassay Stabilizer, and the 5X concentrate, effectively preserves the conformation and activity of dried proteins while simultaneously blocking the surface to reduce non-specific binding. The product can be used to stabilize antibodies, antigens or enzymes on an assortment of immunoassay components including polystyrene plates, tubes, glass, membranes, and filter paper.