Learn More
Surmodics Inc StabilCoat™ Plus Stabilizer, 1L
SDP
Catalog No. 50949951
Encompass_Preferred
StabilCoat™ Plus Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilCoat™ Plus stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.
Catalog No. 50-949-951 Supplier Surmodics Inc Supplier No. SC021000
StabilCoat™ Plus Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilCoat™ Plus stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.